Order Entry
Austria
ContactUsLinkComponent
2-Bromopropane for synthesis, Sigma-Aldrich®
Flammable
Health
2-Bromopropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Health
2-Bromopropane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    3.18 g/l (20 °C)
  • Melting Point:
    -89 °C
  • Molar Mass:
    122.99 g/mol
  • Boiling Point:
    59 - 60 °C
  • Vapor Pressure:
    224 hPa (20 °C)
  • Flash Point:
    19 °C
  • Refractive Index:
    1.4252 (20 °C, 589 nm)
  • Explosion Limit:
    4.6 %(V)
  • Density:
    1.31 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together