Silica gel drying agent, with moisture indicator (orange gel), Supelco®
1.03805.0001
: Merck
:
- Pack type:Metal can
- Pk:10
- Seq:0010
- Environmentally Preferable:
- Solubility:1 g/l
- Melting Point:1000 °C
- Water Absorption:250 g/kg (20 °C)
:
:60.08 g/mol
:M-Clarity™ quality level = MQ200
:Ambient
:63231-67-4
:7631-86-9
:SiO₂
:MFCD00011232