Order Entry
Austria
ContactUsLinkComponent
 
Silica gel sachet: 4 x 7 cm desiccant, with moisture indicator (orange gel), Supelco®
 :  Merck
 
Silica gel sachet: 4 x 7 cm desiccant, with moisture indicator (orange gel), Supelco®
 :  Merck

 

  • Water absorption capacity (24 h, 80 % relative humidity):
    ≥ 20 %
  • Loss on drying (140°C, 3 h):
    ≤ 10 %
  • Water Absorption:
    250 g/kg (20 °C)

 

Frequently Bought Together