Silica gel 1 - 4 mm drying agent, with moisture indicator (brown gel), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together