Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 1.0 N, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid 1.0 N, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Hydrochloric acid 1.0 N, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    0.98 - 1.02
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤3 ppm
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality:
    0.98 - 1.02
  • Ammonium (NH₄):
    ≤ 3 ppm
  • Trace Impurities - Heavy Metals:
    ≤ 0.5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.1 ppm
  • Sulfate (SO₄):
    ≤ 1 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together