Magnesium nitrate hexahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:13446-18-9
:10213-15-7
Formula:Mg(NO₃)₂·6H₂O
MDL Number:MFCD00149779
Boiling Pt:330
Melting Pt:89
Density:1.63 g/cm³ (20 °C)
Molecular Weight:256.41 g/mol
Flash Pt:93.3
Merck Index:13,05697
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:233-826-7
UN:1474
:GHS03
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:420 g/l (20 °C)
- Melting Point:89 °C (for the hexahydrate)
- Molar Mass:256.41 g/mol
- Density:1.46 g/cm3 (20 °C)
- pH:5.0 - 7.0 (50 g/l, H2O, 25 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.05853.0500_TDS_EN ProductPage_105853
Product specification
msds_105853
SDS
Frequently Bought Together