

Calcium nitrate tetrahydrate ≥98.0%, AnalaR NORMAPUR® analytical reagent
:13477-34-4
:
:Ca(NO₃)₂·4H₂O
:MFCD00149604
:236.15 g/mol
:13,01687
:∼42 °C
:1.86 g/cm³ (20 °C)
:Ambient
:233-332-1
:1454
:5.1,III
:GHS07
:GHS05
- Assay:Min. 98.0 %
- Acidity or alkalinity:Max. 0.005 meq/g
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Not precipitated by (COONH4)2 (as SO4):Max. 0.25 %
- Cl (Chloride):Max. 10 ppm
- NH4 (Ammonium):Max. 20 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ba (Barium):Max. 50 ppm
- Fe (Iron):Max. 5 ppm
Frequently Bought Together