Order Entry
Austria
ContactUsLinkComponent
Calcium nitrate tetrahydrate, Suprapur® for trace analysis, Supelco®
Corrosive
Irritant
Calcium nitrate tetrahydrate, Suprapur® for trace analysis, Supelco®
Catalog # 1.02123.0100
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Calcium nitrate tetrahydrate, Suprapur® for trace analysis, Supelco®
Catalog # 1.02123.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    2710 g/l (40 °C)
  • Melting Point:
    45 °C
  • Molar Mass:
    236.15 g/mol
  • Bulk Density:
    1000 kg/m3
  • Density:
    1.82 g/cm3 (20 °C)
  • pH:
    5.0 - 7.0 (50 g/l, H2O, 25 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

About this item

 :
 :236.15 g/mol
Merck Index:13,01687
Melting Pt:∼42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
 :M-Clarity™ quality level = MQ100
 :GHS05
 :GHS07
CAS Number:13477-34-4
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604