Order Entry
Austria
ContactUsLinkComponent
Calcium nitrate tetrahydrate 99-103%, Puriss. p.a. ACS, Fluka™
Irritant
Corrosive
Calcium nitrate tetrahydrate 99-103%, Puriss. p.a. ACS, Fluka™
Certificates
Irritant
Corrosive
Calcium nitrate tetrahydrate 99-103%, Puriss. p.a. ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    99.0 - 103.0 %
  • pH (5 %, 25 °C):
    5.0 - 7.0
  • Water insoluble matter:
    Max. 0.005 %
  • Barium (Ba):
    Max. 0.005 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.01 %
  • Sodium (Na):
    Max. 0.01 %
  • Strontium (Sr):
    Max. 0.01 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Ammonium (NH₄):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.002 %
  • Nitrite (NO₂):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.002 %

Specification Test Results

Related Information

Frequently Bought Together