Order Entry
Austria
ContactUsLinkComponent
Magnesium chloride hexahydrate ≥97%, GPR RECTAPUR®
Magnesium chloride hexahydrate ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Magnesium chloride hexahydrate ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %
  • Alkaline metals (as SO4):
    Max. 2.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Ca (Calcium):
    Max. 0.1 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together