Order Entry
Austria
ContactUsLinkComponent
Magnesium chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Magnesium chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    99.0 - 102.0 %
  • Ammonium (NH4):
    max. 0.002 %
  • Barium (Ba):
    max. 0.005 %
  • Calcium (Ca):
    max. 0.01 %
  • Potassium (K):
    max. 0.005 %
  • Sodium (Na):
    max. 0.005 %
  • Strontium (Sr):
    max. 0.005 %
  • Insoluble Matter:
    max. 0.005 %
  • Nitrate (NO3):
    max. 0.001 %
  • Sulfate (SO4):
    max. 0.002 %
  • Iron (Fe):
    max. 5
  • Manganese (Mn):
    max. 5
  • Heavy Metals (as Pb):
    max. 5
  • Phosphate (PO4):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together