Magnesium chloride hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Magnesium dichloride
CAS Number:7791-18-6
:
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781
Boiling Pt:1412
Melting Pt:117 °C (dec.)
Density:1.570
Molecular Weight:203.3 g/mol
Merck Index:13,05684
Storage Temperature:Ambient
EINECS:232-094-6
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together