
Calcium chloride dihydrate 97.0-103.0% Ph. Eur.
:10035-04-8
:
:CaCl₂·2H₂O
:MFCD00149613
:147.01 g/mol
:13,01660
:1600 °C (1013 hPa)
:176 °C
:1.85 g/cm³ (20 °C)
:233-140-8
:GHS07
:Ambient
- Assay:97.0 - 103.0 %
- Appearance:Conforms (see CoA/CoS)
- Identification (Cl):Passes test
- Identification (Ca):Passes test
- Appearance of solution:Passes test
- Acidity or alkalinity:Passes test
- SO4 (Sulphate):≤300 ppm
- Al (Aluminium):Passes test
- Fe (Iron):≤10 ppm
- Magnesium and alkali metals:≤0.5 %
- Heavy metals (as Pb):≤20 ppm
- Residual solvents:Passes test
- As (Arsenic):≤2 ppm
- Cd (Cadmium):≤1 ppm
- Co (Cobalt):≤1 ppm
- Hg (Mercury):≤2 ppm
- Ni (Nickel):≤1 ppm
- Pb (Lead):≤5 ppm
- V (Vanadium):≤1 ppm
- Conforms to Ph.Eur.:Passes test
10L Plastic bucket for solids UN Y (ZPACK13247010) 1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 50L Plastic bucket UN Y (ZPACK13247050)
Packaging Information
22322_CALCIUM CHLORIDE 2H2O PH.EUR_ALLERGEN_Rev1 Allergen_VWRC_22322_CALCIUM CHLORIDE 2H2O PH.EUR.pdf
ALLERGEN
22322_CALCIUM CHLORIDE 2H2O PH.EUR_BSE_REV1
BSE/TSE Certificates
Calcium Chloride Dihydrate 97 - 103% Ph. Eur.
Additional Product Details
spec_VWRC22322.295 spec_VWRC22322.295 spec_VWRC22322.364 spec_VWRC22322.364 spec_VWRC22322.466
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together