
Calcium chloride dihydrate 99.0-105.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10035-04-8
:
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613
:147.01 g/mol
Merck Index:14,1659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:0.835
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together