Order Entry
Austria
ContactUsLinkComponent
 
 
Sodium tungstate dihydrate, EMSURE® for analysis, Supelco®
 :  Merck
 
undefined
 
Sodium tungstate dihydrate, EMSURE® for analysis, Supelco®
 :  Merck

 

  • Solubility:
    730 g/l (20 °C)
  • Melting Point:
    100 °C Elimination of water of crystallisation
  • Molar Mass:
    329.86 g/mol
  • Bulk Density:
    1600 kg/m3
  • Density:
    3.23 g/cm3 (20 °C)
  • pH:
    9 - 11.5 (100 g/l, H2O, 20 °C)

 

Frequently Bought Together