Sodium tungstate dihydrate 99+% ACS
: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate
:10213-10-2
:
:Na₂WO₄·2H₂O
:MFCD00149190
:329.85 g/mol
:12,08847
:100 °C
:3,23 g/cm³ (20 °C)
:236-743-4
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White to light yellow Crystalline powder or flakes
- Gravimetry:99.0 to 101.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Molybdenum (Mo):≤10 ppm
- Heavy metals & Fe (as Pb):≤0.001 %
- Titrable free base:≤0.02 meq/g
- Insoluble matter:≤0.01 %
Frequently Bought Together