Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate pentahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Sodium thiosulphate pentahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Sodium thiosulphate pentahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    701 g/l (20 °C)
  • Melting Point:
    48 °C
  • Molar Mass:
    248.21 g/mol
  • Bulk Density:
    1000 kg/m3
  • Density:
    1.74 g/cm3 (20 °C)
  • pH:
    6.0 - 7.5 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together