Order Entry
Austria
ContactUsLinkComponent
 
Sodium thiosulphate pentahydrate 99.0-101.0% (by iodometry), crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
 
Sodium thiosulphate pentahydrate 99.0-101.0% (by iodometry), crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
 

 

  • Assay (iodometric):
    99.0 - 101.0 %
  • Assay (iodometric, calc.on anhydrous substance):
    99.0 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • pH-value (10 %; water):
    6.0 - 8.4
  • Sulfate and sulphite (as SO₄):
    ≤ 0.2 %
  • Sulfide (S):
    passes test
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • Ca (Calcium):
    passes test
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Water:
    32.0 - 37.0 %

 

 

Frequently Bought Together