Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate pentahydrate, J.T.Baker®
Sodium thiosulphate pentahydrate, J.T.Baker®
Catalog # 1779.9050
Supplier:  Avantor
CAS Number:  
Sodium thiosulphate pentahydrate, J.T.Baker®
Catalog # 1779.9050
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
 :248.19 g/mol
Merck Index:12,08844
Melting Pt:48 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186