Sodium dihydrogen phosphate monohydrate, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:10049-21-5
:
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208
:137.99 g/mol
Merck Index:13,08734
Melting Pt:100 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
Frequently Bought Together