Order Entry
Austria
Orders LinkContactUsLinkComponent
Disodium hydrogen phosphate dihydrate

Disodium hydrogen phosphate dihydrate

Marketsource
Supplier:   Roth Carl
Disodium hydrogen phosphate dihydrate
Supplier:  Roth Carl
Marketsource
Certificates
Disodium hydrogen phosphate dihydrate

3 Options Available Below

Product Details & Documents
Synonyms:  sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt
  • CAS Number:10028-24-7
  • Formula:Na₂HPO₄·2H₂O
  • MDL Number:MFCD00149182
  • Molecular Weight:177.99 g/mol
  • Melting Pt:92.5 °C
  • Density:2.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-448-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 32,70 €
    32,70 €
    1 * 500 g
       32,70 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 54,50 €
    54,50 €
    1 * 1 kg
       54,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 109,00 €
    109,00 €
    1 * 2,5 kg
       109,00 €

    Specifications:
    Marketsource
    Marketsource