Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dihydrate ≥99.0% (by acidimetry), LiChropur™ buffer substance, for chromatography, Supelco®
Disodium hydrogen phosphate dihydrate ≥99.0% (by acidimetry), LiChropur™ buffer substance, for chromatography, Supelco®
Catalog # 1.19753.0250
Supplier:  Merck
CAS Number:  
Disodium hydrogen phosphate dihydrate ≥99.0% (by acidimetry), LiChropur™ buffer substance, for chromatography, Supelco®
Catalog # 1.19753.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Identity:
    passes test
  • pH-value (5 %; water):
    9.0 - 9.2
  • Assay (acidimetric):
    ≥ 99.0 %
  • Self-absorption (215 nm; 0.2 mol/l; 1 cm; water):
    ≤ 0.15
  • Self-absorption (220 nm; 0.2 mol/l; 1 cm; water):
    ≤ 0.05
  • Self-absorption (254 nm; 0.2 mol/l; 1 cm; water):
    ≤ 0.02

Specification Test Results

About this item

 :
 :177.99 g/mol
Melting Pt:92.5 °C
Density:2.1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:10028-24-7
Formula:Na₂HPO₄·2H₂O
MDL Number:MFCD00149182