Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis, Supelco®
Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric):
    ≥ 99.5 %
  • Identity:
    passes test
  • pH-value (5 %; water):
    9.0 - 9.2
  • Chloride (Cl):
    ≤ 0.001 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Total nitrogen (N):
    ≤ 0.001 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • Cu (Copper):
    ≤ 0.0003 %
  • Fe (Iron):
    ≤ 0.001 %
  • K (Potassium):
    ≤ 0.005 %
  • Pb (Lead):
    ≤ 0.001 %

Specification Test Results

Related Information

Frequently Bought Together