Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™
Disodium hydrogen phosphate dodecahydrate ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™
Lieferant:  Honeywell Chemicals
Zertifikate
Disodium hydrogen phosphate dodecahydrate ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™
Lieferant:  Honeywell Chemicals

Spezifikation Testergebnisse

  • Assay titr.:
    99.0 - 101.0 %
  • pH (5 %, 25 °C):
    8.8 - 9.4
  • Arsenic (As):
    Max. 0.00005 %
  • Calcium (Ca):
    Max. 0.001 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Cobalt (Co):
    Max. 0.0005 %
  • Chromium (Cr):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0005 %
  • Potassium (K):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Magnesium (Mg):
    Max. 0.0005 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Fluoride (F):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Appearance:
    complying
  • Solution 0.5 g at 10 mL (H₂O):
    complying
  • Appearance of the solution:
    complying
  • Colour of the solution:
    complying
  • Turbidity of solution:
    Max. 3.5 NTU

Spezifikation Testergebnisse

Zugehörige Informationen

Frequently Bought Together