Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate ≥99%, Gen-Apex for molecular biology
Disodium hydrogen phosphate dodecahydrate ≥99%, Gen-Apex for molecular biology
Catalog # 33617.265
Supplier:  VWR Chemicals
CAS Number:  
undefined
Disodium hydrogen phosphate dodecahydrate ≥99%, Gen-Apex for molecular biology
Catalog # 33617.265
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Colouration (0,1 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Transmittance (230 nm):
    Min. 90 %
  • Transmittance (260 nm):
    Min. 95 %
  • Transmittance (280 nm):
    Min. 97 %
  • Transmittance (320 nm):
    Min. 99 %

Specification Test Results

About this item

 :
 :358.14 g/mol
Merck Index:13,08733
Melting Pt:35 °C
Density:1.52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:10039-32-4
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181