Order Entry
Austria
ContactUsLinkComponent
 
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
 
undefined
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
 

 

  • Assay (calculated on anhydrous):
    Min. 99.0 %
  • pH (20°C; 5 %):
    9.0 - 9.4
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 0.2 %
  • Loss on drying (120°C):
    50.0 - 60.5 %
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • As (Arsenic):
    Max. 0.5 ppm
  • Cu (Copper):
    Max. 2 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 100 ppm
  • Pb (Lead):
    Max. 5 ppm
  • Zn (Zinc):
    Max. 1 ppm

 

Frequently Bought Together