Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate ≥98%, purified
Disodium hydrogen phosphate dodecahydrate ≥98%, purified
Supplier:  VWR Chemicals
Certificates
undefined
Disodium hydrogen phosphate dodecahydrate ≥98%, purified
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.05 %
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together