Order Entry
Austria
ContactUsLinkComponent
Trisodium phosphate dodecahydrate ≥98.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Irritant
Trisodium phosphate dodecahydrate ≥98.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Catalog # 71911-1KG
CAS Number:  
Irritant
Trisodium phosphate dodecahydrate ≥98.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Catalog # 71911-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Free alkali (as NaOH):
    Max. 2.5 %
  • Insoluble matter:
    Max. 0.01 %
  • Arsenic (As):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Total nitrogen (as N):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :380.12 g/mol
Merck Index:13,08736
Melting Pt:75 °C (dec.)
Density:1,62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-509-8
ADR:8,III
REACH:01-2119489800-32-xxxx
 :GHS07
CAS Number:10101-89-0
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198