Order Entry
Austria
ContactUsLinkComponent
 
Sodium molybdate(VI) dihydrate
Catalog # MOLE11137082-1KG
Supplier:  Molekula
CAS Number:  
Sodium molybdate(VI) dihydrate
Catalog # MOLE11137082-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-551-7
CAS Number:10102-40-6
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170