

di-Sodium tetraborate decahydrate ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1303-96-4
:
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193
:381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
:GHS07
Specification Test Results
- Solubility:51.4 g/l (20 °C)
- Melting Point:75 °C Elimination of water of crystallisation
- Molar Mass:381.32 g/mol
- Bulk Density:750 kg/m3
- Vapor Pressure:0.213 hPa (20 °C)
- Density:1.72 g/cm3 (20 °C)
- pH:9.2 (47 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06308.0500_TDS_EN MERC1.06308.1000_TDS ProductPage_106308
Product specification
msds_106308
SDS
Frequently Bought Together