Order Entry
Austria
ContactUsLinkComponent
 
 
di-Sodium tetraborate decahydrate ≥99.5%, Puriss. p.a. ACS, ISO buffer substance, Fluka™
 
 
di-Sodium tetraborate decahydrate ≥99.5%, Puriss. p.a. ACS, ISO buffer substance, Fluka™
 

 

  • Assay:
    99.5 - 105.0 %
  • Water insoluble matter:
    Max. 0.001 %
  • pH (5 %, 20°C):
    9.0 - 9.6
  • pH (0.01 m, 25°C):
    9.15 - 9.20
  • Arsenic (As):
    Max. 0.0001 %
  • Calcium (Ca):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %

 

 

Frequently Bought Together