Order Entry
Austria
ContactUsLinkComponent
Silver nitrate 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Environment
Silver nitrate 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7246.1000
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7246.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (mol/l):
    0.995 - 1.005

Specification Test Results

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,II
 :GHS05
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414