Silver nitrate 0,1 mol/l (0,1 N) in aqueous solution, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:231-853-9
UN:1760
:GHS05
:GHS09
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Related Information
Frequently Bought Together