ContactUsLinkComponent
Silver nitrate standard solution 0.1 mol/l (0.1 N) in water
Corrosive
Environment
Silver nitrate standard solution 0.1 mol/l (0.1 N) in water
Certificates
undefined
Corrosive
Environment
Silver nitrate standard solution 0.1 mol/l (0.1 N) in water
Certificates
Specification Test Results
  • Appearance:
    Clear colorless Solution
  • Strength:
    0.0998 to 0.1002 N (20°C)