Order Entry
Austria
ContactUsLinkComponent
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7245.1000
Supplier:  Avantor
CAS Number:  
undefined
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7245.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0495 - 0.0505

Specification Test Results

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414