Order Entry
Austria
ContactUsLinkComponent
Magnesium chloride 2 mol/l (4 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL68475-100ML-F
Supplier:  Merck
CAS Number:  
Magnesium chloride 2 mol/l (4 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL68475-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

 :
 :95.21 g/mol
Boiling Pt:1412
Melting Pt:715
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106
Merck Index:13,05684