ContactUsLinkComponent
Sulphuric acid 0.05 M H2SO4 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Corrosive
Sulphuric acid 0.05 M H2SO4 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Certificates
Corrosive
Sulphuric acid 0.05 M H2SO4 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Certificates
Specification Test Results
  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective