ContactUsLinkComponent
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Sulphuric acid 0.02 mol/l (0.04 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.01996 - 0.02004 mol/l
  • NIST traceable:
    Confirmed