Order Entry
Austria
ContactUsLinkComponent
 
Sulphuric acid 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
  95032.1000
 :  
undefined
Sulphuric acid 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
  95032.1000
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Titer:
    0.00995 - 0.01005 mol/l

 

 

 :
 :98.08 g/mol
 :1.01 g/cm³ (20 °C)
 :Ambient
 :231-639-5
 :2796
 :8,II
 :7664-93-9
 :H₂SO₄
 :MFCD00064589