Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30147.408
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30147.408
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.00499 - 0.00501 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589