Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium thiosulphate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31547.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium thiosulphate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31547.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499