Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate 0.01 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Sodium thiosulphate 0.01 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09909.0001
Supplier:  Merck
CAS Number:  
Sodium thiosulphate 0.01 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09909.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.02 g/cm3 (20 °C)
  • pH:
    9 - 10 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :158.11 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499