
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:1.078 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- NIST traceable:Confirmed
- Titer (20°C; real value 0.2 % accuracy):1.996 - 2.004 mol/l
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC98108.290 spec_VWRC98108.290 spec_VWRC98108.360 spec_VWRC98108.360 spec_VWRC98108.400
Product specification
More
Frequently Bought Together