ContactUsLinkComponent
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Corrosive
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier:  Merck
Certificates
Corrosive
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier:  Merck
Certificates
Specification Test Results
  • Form:
    liquid
  • Amount-of-substance concentration:
    1.990 - 2.010
  • Measurement uncertainty:
    +/- 0.006
  • Traceability:
    NIST SRM