Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7036.1000
Supplier:  Avantor
CAS Number:  
Sodium hydroxide 2 mol/l (2 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7036.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Titer (mol/l):
    1.995 - 2.005
  • Chloride (Cl):
    max. 5
  • Iron (Fe):
    max. 0.5
  • Heavy Metals (as Pb):
    max. 1

Specification Test Results

About this item

Synonyms: Caustic soda
 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548