
Sodium hydroxide 10 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:109 °C (1013 hPa)
:1.33 g/cm3 (20 °C)
:Refrigerator
:215-185-5
:1824
:8,II
:Filtered through a 0.2 µm filter.
:GHS05
- Assay (acidimetric,NaOH):9.9 - 10.1 mol/l
- Assay (acidimetric):29.9 - 30.3 %
- Identity:passes test
- Carbonate (as Na₂CO₃):≤ 1.0 %
- Chloride (Cl):≤ 20 ppm
- Sulfate (SO₄):≤ 30 ppm
- Heavy metals (as Pb):≤ 5.0 ppm
- Al (Aluminium):≤ 10 ppm
- Fe (Iron):≤ 3 ppm
- Ni (Nickel) *:≤ 2.00 ppm
- Residual solvents (ICH Q3C):...excluded by production process
- Endotoxins:≤ 2.0 I.U./ml
Frequently Bought Together