ContactUsLinkComponent
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
 :  Merck
undefined
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
 :  Merck
  • Form:
    liquid
  • Amount-of-substance concentration:
    0.4975 - 0.5025
  • Measurement uncertainty:
    +/- 0.0015
  • Traceability:
    NIST SRM
Frequently Bought Together