0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution
PRLSMC309338.6010
:
PROLABO CMP
:
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution
PRLSMC309338.6010
:
PROLABO CMP
:
Pk:
10 L
Seq:
5214
:
:
40 g/mol
:
∼105 °C (1013 hPa)
:
1.02 g/cm³ (25 °C)
:
Ambient
:
215-185-5
:
1824
:
8,III
:
GHS07
:
1310-73-2
:
NaOH
:
MFCD00003548