
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
: Merck
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:∼105 °C (1013 hPa)
:1.02 g/cm³ (25 °C)
:Ambient
:215-185-5
:1824
:8,II
:M-Clarity™ quality level = MQ100
:GHS05
- Form:liquid
- Amount-of-substance concentration:0.24875 - 0.25125
- Measurement uncertainty:0.00075
- Traceability:NIST SRM
Frequently Bought Together