Cover glasses
Supplier: Brand
Certificates
About this item
These cover glasses are made of pure white clear glass and offer good chemical resistance.
- Automatic production guarantees ready to use quality
- Clean, dust and grease free
- Distortion-free
Flatness: ±3 µm
Frequently Bought Together